Assay Details
| Gene Symbol: Larp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024526.2 | NP_001019697.2 | 13 - 14 | 1716 | 98 | |
| NM_001080948.2 | NP_001074417.1 | 13 - 14 | 1729 | 98 | ||
| NM_001284521.1 | NP_001271450.1 | 13 - 14 | 1703 | 98 | ||
| NM_001284522.1 | NP_001271451.1 | 13 - 14 | 1726 | 98 | ||
| NM_001284523.1 | NP_001271452.1 | 13 - 14 | 1719 | 98 | ||
| GenBank mRNA | AK049756.1 | - | 13 - 14 | 1643 | 98 | |
| AK139636.1 | - | 13 - 14 | 1724 | 98 | ||
| AK139909.1 | - | 13 - 14 | 1567 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Larp4
Genomic Map