Assay Details
| Gene Symbol: Larp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024526.2 | NP_001019697.2 | 4 - 5 | 571 | 139 | |
| NM_001080948.2 | NP_001074417.1 | 4 - 5 | 584 | 139 | ||
| NM_001284521.1 | NP_001271450.1 | 4 - 5 | 558 | 139 | ||
| NM_001284522.1 | NP_001271451.1 | 4 - 5 | 581 | 139 | ||
| NM_001284523.1 | NP_001271452.1 | 4 - 5 | 574 | 139 | ||
| GenBank mRNA | AK049756.1 | - | 4 - 5 | 498 | 139 | |
| AK139636.1 | - | 4 - 5 | 577 | 139 | ||
| AK139909.1 | - | 4 - 5 | 422 | 139 | ||
| AK165392.1 | - | 4 - 5 | 519 | 139 | ||
| AK172198.1 | - | 4 - 5 | 516 | 139 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Larp4
Genomic Map