Assay Details
| Gene Symbol: Tada1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030245.3 | NP_084521.1 | 4 - 5 | 561 | 96 | |
| GenBank mRNA | AK013599.1 | - | 4 - 5 | 328 | 96 | |
| AK151132.1 | - | 4 - 5 | 423 | 96 | ||
| BC002307.1 | - | 4 - 5 | 363 | 3592046 | 96 | |
| BC027337.1 | - | 4 - 5 | 428 | 3995355 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tada1
Genomic Map