Assay Details
| Gene Symbol: Cacna1i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044308.2 | NP_001037773.2 | 6 - 7 | 1293 | 83 | |
| GenBank mRNA | AK147585.1 | - | 6 - 7 | 1293 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cacna1i
Genomic Map