Assay Details
| Gene Symbol: Cuta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026307.3 | NP_080583.3 | 4 - 5 | 401 | 76 | |
| NM_026948.3 | NP_081224.1 | 3 - 4 | 349 | 76 | ||
| GenBank mRNA | AK002828.1 | - | 4 - 5 | 345 | 76 | |
| AK003381.1 | - | 3 - 4 | 289 | 76 | ||
| AK003647.1 | - | 4 - 5 | 343 | 76 | ||
| AK004197.1 | - | 4 - 5 | 343 | 76 | ||
| AK159336.1 | - | 4 - 5 | 348 | 76 | ||
| BC021463.1 | - | 2 - 3 | 463 | 76 | ||
| BC024422.1 | - | 4 - 5 | 331 | 5005082 | 76 | |
| BC037500.1 | - | 2 - 3 | 327 | 76 | ||
| CA323205.1 | - | 3 - 4 | 315 | 76 | ||
| CK624897.1 | - | 3 - 4 | 326 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cuta
Genomic Map