Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500016505.mm.1 |
NM_001013022.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016505.mm.1 |
NM_001013022.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016505.mm.1 |
TC1500002053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016507.mm.1 |
NM_001013022.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016507.mm.1 |
NM_001013022.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016507.mm.1 |
TC1500002053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016504.mm.1 |
NM_001013022.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016504.mm.1 |
NM_001013022.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016504.mm.1 |
TC1500002053.mm.1 |
MTA TC |