Assay Details
| Gene Symbol: Tbc1d22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145476.2 | NP_663451.2 | 2 - 3 | 234 | 71 | |
| GenBank mRNA | AK039627.1 | - | 2 - 3 | 247 | 71 | |
| AK146872.1 | - | 2 - 3 | 235 | 71 | ||
| AK154226.1 | - | 2 - 3 | 256 | 71 | ||
| AK155459.1 | - | 2 - 3 | 248 | 71 | ||
| AK155770.1 | - | 2 - 3 | 277 | 71 | ||
| BC066009.1 | - | 2 - 3 | 234 | 5708486 | 71 | |
More Information
Set Membership: |
|

Tbc1d22a
Genomic Map