Assay Details
| Gene Symbol: Tbc1d22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145476.2 | NP_663451.2 | 4 - 5 | 753 | 72 | |
| GenBank mRNA | AK039627.1 | - | 4 - 5 | 766 | 72 | |
| AK146872.1 | - | 4 - 5 | 754 | 72 | ||
| AK154226.1 | - | 4 - 5 | 775 | 72 | ||
| AK154602.1 | - | 4 - 5 | 796 | 72 | ||
| AK155459.1 | - | 4 - 5 | 767 | 72 | ||
| AK155770.1 | - | 4 - 5 | 796 | 72 | ||
| BC023106.1 | - | 1 - 2 | 148 | 4016550 | 72 | |
| BC066009.1 | - | 4 - 5 | 753 | 5708486 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023106.1 transcripts.
Set Membership: |
|

Tbc1d22a
Genomic Map