Assay Details
| Gene Symbol: Tbc1d22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145476.2 | NP_663451.2 | 5 - 6 | 817 | 63 | |
| GenBank mRNA | AK039627.1 | - | 5 - 6 | 830 | 63 | |
| AK146872.1 | - | 5 - 6 | 818 | 63 | ||
| AK154226.1 | - | 5 - 6 | 839 | 63 | ||
| AK154602.1 | - | 5 - 6 | 860 | 63 | ||
| AK155459.1 | - | 5 - 6 | 831 | 63 | ||
| AK155770.1 | - | 5 - 6 | 860 | 63 | ||
| AK194823.1 | - | 1 - 2 | 42 | 63 | ||
| AK194954.1 | - | 226 | 63 | |||
| AK208524.1 | - | 1 - 2 | 42 | 63 | ||
| AK214247.1 | - | 1 - 2 | 42 | 63 | ||
| AK214703.1 | - | 1 - 2 | 42 | 63 | ||
| AK216899.1 | - | 1 - 2 | 42 | 63 | ||
| AK216948.1 | - | 1 - 2 | 42 | 63 | ||
| AK217113.1 | - | 1 - 2 | 42 | 63 | ||
| BC023106.1 | - | 2 - 3 | 212 | 4016550 | 63 | |
| BC066009.1 | - | 5 - 6 | 817 | 5708486 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023106.1 transcripts.
Set Membership: |
|

Tbc1d22a
Genomic Map