Assay Details
| Gene Symbol: Gtse1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168672.1 | NP_001162143.1 | 9 - 10 | 1835 | 131 | |
| NM_013882.2 | NP_038910.1 | 8 - 9 | 1805 | 131 | ||
| GenBank mRNA | AJ222580.1 | - | 8 - 9 | 1775 | 131 | |
| AK081582.1 | - | 9 - 10 | 1798 | 131 | ||
More Information
Set Membership: |
|

Gtse1
Genomic Map