Assay Details
| Gene Symbol: Tbkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198100.2 | NP_932768.2 | 6 - 7 | 1001 | 92 | |
| GenBank mRNA | BC057590.1 | - | 6 - 7 | 1001 | 6810986 | 92 |
More Information
Updated Mapping Information
Set Membership: |
|

Tbkbp1
Genomic Map