| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700004026.mm.1 |
TC1700000472.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004028.mm.1 |
NM_172617.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004028.mm.1 |
NM_172617.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004028.mm.1 |
TC1700000472.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004029.mm.1 |
NM_172617.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004029.mm.1 |
NM_172617.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004029.mm.1 |
TC1700000472.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004034.mm.1 |
NM_172617.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004034.mm.1 |
NM_172617.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004034.mm.1 |
TC1700000472.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004031.mm.1 |
NM_172617.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004031.mm.1 |
NM_172617.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004031.mm.1 |
TC1700000472.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004025.mm.1 |
TC1700000472.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004033.mm.1 |
NM_172617.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004033.mm.1 |
NM_172617.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004033.mm.1 |
TC1700000472.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004038.mm.1 |
NM_172617.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004038.mm.1 |
NM_172617.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004038.mm.1 |
TC1700000472.mm.1 |
MTA TC |