Assay Details
| Gene Symbol: Trim23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029533.1 | - | 1 - 2 | 126 | 122 | |
| AK030843.1 | - | 1 - 2 | 126 | 122 | ||
| AK036210.1 | - | 1 - 2 | 127 | 123 | ||
| AK039280.1 | - | 1 - 2 | 138 | 122 | ||
| AK042070.1 | - | 1 - 2 | 129 | 122 | ||
| AK088922.1 | - | 1 - 2 | 126 | 122 | ||
| BC056390.1 | - | 1 - 2 | 119 | 6854944 | 122 | |
| BC059017.1 | - | 1 - 2 | 119 | 6848690 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim23
Genomic Map