Assay Details
| Gene Symbol: Trim23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030731.3 | NP_109656.1 | 4 - 5 | 2721 | 87 | |
| GenBank mRNA | AK029533.1 | - | 5 - 6 | 869 | 87 | |
| AK030843.1 | - | 5 - 6 | 869 | 87 | ||
| AK030862.1 | - | 4 - 5 | 2722 | 87 | ||
| AK036210.1 | - | 5 - 6 | 870 | 87 | ||
| AK039280.1 | - | 5 - 6 | 881 | 87 | ||
| AK042070.1 | - | 5 - 6 | 872 | 87 | ||
| BC056390.1 | - | 5 - 6 | 862 | 6854944 | 87 | |
| BC059017.1 | - | 5 - 6 | 862 | 6848690 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim23
Genomic Map