Assay Details
| Gene Symbol: Dnajc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030046.2 | NP_084322.2 | 4 - 5 | 590 | 86 | |
| GenBank mRNA | AK140224.1 | - | 4 - 5 | 591 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc21
Genomic Map