Assay Details
| Gene Symbol: Arfgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025445.4 | NP_079721.2 | 14 - 15 | 1531 | 78 | |
| GenBank mRNA | AK007732.1 | - | 14 - 15 | 1451 | 78 | |
| AK028990.1 | - | 14 - 15 | 1532 | 78 | ||
| AK075788.1 | - | 14 - 15 | 1517 | 78 | ||
| AK169107.1 | - | 14 - 15 | 1438 | 78 | ||
| BC004081.1 | - | 14 - 15 | 1467 | 3592719 | 78 | |
| BC060369.1 | - | 14 - 15 | 1455 | 6310619 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arfgap3
Genomic Map