Assay Details
| Gene Symbol: Arfgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025445.4 | NP_079721.2 | 5 - 6 | 578 | 108 | |
| GenBank mRNA | AK007732.1 | - | 5 - 6 | 495 | 108 | |
| AK028990.1 | - | 5 - 6 | 579 | 108 | ||
| AK075788.1 | - | 5 - 6 | 567 | 108 | ||
| AK169107.1 | - | 5 - 6 | 488 | 108 | ||
| BC004081.1 | - | 5 - 6 | 514 | 3592719 | 108 | |
| BC060369.1 | - | 5 - 6 | 505 | 6310619 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arfgap3
Genomic Map