Assay Details
| Gene Symbol: Arfgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025445.4 | NP_079721.2 | 8 - 9 | 775 | 87 | |
| GenBank mRNA | AK028990.1 | - | 8 - 9 | 776 | 87 | |
| AK075788.1 | - | 8 - 9 | 764 | 87 | ||
| AK169107.1 | - | 8 - 9 | 685 | 87 | ||
| AK215301.1 | - | 3 - 4 | 152 | 87 | ||
| BC004081.1 | - | 8 - 9 | 711 | 3592719 | 87 | |
| BC060369.1 | - | 8 - 9 | 702 | 6310619 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arfgap3
Genomic Map