Assay Details
| Gene Symbol: Sun2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205345.1 | NP_001192274.1 | 11 - 12 | 1471 | 76 | |
| NM_001205346.1 | NP_001192275.1 | 11 - 12 | 1465 | 76 | ||
| NM_194342.3 | NP_919323.2 | 10 - 11 | 1375 | 76 | ||
| GenBank mRNA | AK156246.1 | - | 11 - 12 | 1472 | 76 | |
| AK171058.1 | - | 11 - 12 | 1434 | 76 | ||
| AK172398.1 | - | 11 - 12 | 1473 | 76 | ||
| AY682987.1 | - | 10 - 11 | 1231 | 76 | ||
| BC098208.1 | - | 11 - 12 | 1385 | 6827666 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sun2
Genomic Map