| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500014787.mm.1 |
NM_001205345.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014787.mm.1 |
NM_001205345.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014787.mm.1 |
NM_001205346.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014787.mm.1 |
NM_001205346.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014787.mm.1 |
NM_194342.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014787.mm.1 |
NM_194342.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014787.mm.1 |
TC1500001885.mm.1 |
MTA TC |