Assay Details
| Gene Symbol: Sun2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205345.1 | NP_001192274.1 | 15 - 16 | 2059 | 60 | |
| NM_001205346.1 | NP_001192275.1 | 15 - 16 | 2053 | 60 | ||
| NM_194342.3 | NP_919323.2 | 14 - 15 | 1963 | 60 | ||
| GenBank mRNA | AK128958.1 | - | 14 - 15 | 1957 | 60 | |
| AK156246.1 | - | 15 - 16 | 2060 | 60 | ||
| AK171058.1 | - | 15 - 16 | 2022 | 60 | ||
| AK172398.1 | - | 15 - 16 | 2061 | 60 | ||
| AK172996.1 | - | 13 - 14 | 2663 | 60 | ||
| AY682987.1 | - | 14 - 15 | 1819 | 60 | ||
| BC023965.1 | - | 4 - 5 | 611 | 60 | ||
| BC098208.1 | - | 14 - 15 | 2304 | 6827666 | 60 | |
| BQ879674.1 | - | 2 - 3 | 175 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sun2
Genomic Map