Assay Details
| Gene Symbol: Sun2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205345.1 | NP_001192274.1 | 7 - 8 | 964 | 74 | |
| NM_001205346.1 | NP_001192275.1 | 7 - 8 | 958 | 74 | ||
| NM_194342.3 | NP_919323.2 | 6 - 7 | 868 | 74 | ||
| GenBank mRNA | AK046318.1 | - | 6 - 7 | 837 | 74 | |
| AK128958.1 | - | 6 - 7 | 862 | 74 | ||
| AK156246.1 | - | 7 - 8 | 965 | 74 | ||
| AK171058.1 | - | 7 - 8 | 927 | 74 | ||
| AK172398.1 | - | 7 - 8 | 965 | 74 | ||
| AK172996.1 | - | 7 - 8 | 789 | 74 | ||
| AY682987.1 | - | 6 - 7 | 724 | 74 | ||
| BC098208.1 | - | 7 - 8 | 878 | 6827666 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sun2
Genomic Map