Assay Details
| Gene Symbol: Trpm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134252.3 | NP_599013.1 | 24 - 25 | 3410 | 67 | |
| GenBank mRNA | AF481480.2 | - | 3679 | 67 | ||
| AY095352.1 | - | 22 - 23 | 3244 | 67 | ||
| BC117934.1 | - | 24 - 25 | 3410 | 40097778 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpm8
Genomic Map