Assay Details
| Gene Symbol: Igfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177642.3 | NP_808310.2 | 16 - 17 | 6283 | 72 | |
| GenBank mRNA | EU429481.1 | - | 2 - 3 | 3155 | 72 | |
| EU429482.1 | - | 1 - 2 | 87 | 72 | ||
| EU429483.2 | - | 11 - 12 | 1436 | 72 | ||
| GU290042.1 | - | 16 - 17 | 6283 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the EU429482.1 transcripts.
Set Membership: |
|

Igfn1
Genomic Map