Assay Details
| Gene Symbol: Igfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177642.3 | NP_808310.2 | 18 - 19 | 6715 | 84 | |
| GenBank mRNA | EU429481.1 | - | 4 - 5 | 3587 | 84 | |
| EU429483.2 | - | 13 - 14 | 1868 | 84 | ||
| GU290042.1 | - | 18 - 19 | 6715 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igfn1
Genomic Map