Assay Details
| Gene Symbol: Lax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159649.1 | NP_001153121.1 | 2 - 3 | 487 | 98 | |
| NM_172842.3 | NP_766430.3 | 2 - 3 | 389 | 98 | ||
| GenBank mRNA | AK040834.1 | - | 2 - 3 | 297 | 98 | |
| AK088503.1 | - | 2 - 3 | 240 | 98 | ||
| AY090785.1 | - | 1 - 2 | 115 | 98 | ||
| BC042765.1 | - | 2 - 3 | 389 | 4188712 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lax1
Genomic Map