Assay Details
| Gene Symbol: Lax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159649.1 | NP_001153121.1 | 3 - 4 | 64 | ||
| NM_172842.3 | NP_766430.3 | 3 - 4 | 64 | |||
| GenBank mRNA | AK088503.1 | - | 3 - 4 | 343 | 64 | |
| AY090785.1 | - | 2 - 3 | 64 | |||
| BC042765.1 | - | 3 - 4 | 492 | 4188712 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lax1
Genomic Map