Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700002776.mm.1 |
NM_024284.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002776.mm.1 |
NM_024284.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002776.mm.1 |
TC1700000354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002777.mm.1 |
NM_024284.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002777.mm.1 |
NM_024284.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002777.mm.1 |
TC1700000354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002780.mm.1 |
NM_024284.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002780.mm.1 |
NM_024284.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002780.mm.1 |
TC1700000354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002786.mm.1 |
NM_001159626.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002786.mm.1 |
NM_001159626.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002786.mm.1 |
NM_024284.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002786.mm.1 |
NM_024284.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002786.mm.1 |
TC1700000354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700002785.mm.1 |
NM_001159626.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002785.mm.1 |
NM_001159626.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002785.mm.1 |
NM_024284.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002785.mm.1 |
NM_024284.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700002785.mm.1 |
TC1700000354.mm.1 |
MTA TC |