Assay Details
| Gene Symbol: Abca17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031621.2 | NP_001026792.2 | 9 - 10 | 1454 | 135 | |
| GenBank mRNA | AB112584.1 | - | 9 - 10 | 1454 | 135 | |
More Information
Set Membership: |
|

Abca17
Genomic Map