Assay Details
| Gene Symbol: Rab11fip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162868.1 | NP_001156340.1 | 2 - 3 | 1689 | 88 | |
| NM_001162869.1 | NP_001156341.1 | 2 - 3 | 1689 | 88 | ||
| GenBank mRNA | AB093257.1 | - | 2 - 3 | 1130 | 88 | |
| AK083658.1 | - | 2 - 3 | 1373 | 88 | ||
More Information
Set Membership: |
|

Rab11fip3
Genomic Map