Assay Details
| Gene Symbol: Krt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033073.3 | NP_149064.1 | 4 - 5 | 749 | 96 | |
| GenBank mRNA | AF509888.1 | - | 4 - 5 | 726 | 96 | |
| AK002434.1 | - | 4 - 5 | 727 | 96 | ||
| AK137217.1 | - | 727 | 96 | |||
| AK146914.1 | - | 4 - 5 | 727 | 96 | ||
| BC010337.1 | - | 3 - 4 | 2743 | 96 | ||
| BC030403.1 | - | 4 - 5 | 760 | 5355662 | 96 | |
More Information
Set Membership: |
|

Krt7
Genomic Map