Assay Details
| Gene Symbol: Slc4a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021530.2 | NP_067505.2 | 19 - 20 | 2874 | 77 | |
| GenBank mRNA | AF224508.1 | - | 19 - 20 | 2820 | 77 | |
| AK034237.1 | - | 19 - 20 | 2876 | 77 | ||
| AK039404.1 | - | 19 - 20 | 2839 | 77 | ||
| AK047863.1 | - | 12 - 13 | 1808 | 77 | ||
| AK171610.1 | - | 18 - 19 | 2650 | 77 | ||
| AK173014.1 | - | 19 - 20 | 2708 | 77 | ||
| BC030388.1 | - | 19 - 20 | 2836 | 5401143 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a8
Genomic Map