Assay Details
| Gene Symbol: Dnase1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025718.3 | NP_079994.2 | 3 - 4 | 347 | 66 | |
| GenBank mRNA | AK003619.1 | - | 3 - 4 | 348 | 66 | |
| AK014633.1 | - | 2 - 3 | 539 | 66 | ||
| BC106188.1 | - | 3 - 4 | 311 | 30135288 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnase1l2
Genomic Map