Assay Details
| Gene Symbol: E4f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF126967.1 | - | 6 - 7 | 899 | 60 | |
| AK033285.1 | - | 6 - 7 | 911 | 60 | ||
| AK077014.1 | - | 960 | 60 | |||
| AK144176.1 | - | 6 - 7 | 856 | 60 | ||
| BC005634.1 | - | 6 - 7 | 830 | 60 | ||
| BC031757.1 | - | 5 - 6 | 1040 | 60 | ||
| BC046459.1 | - | 6 - 7 | 969 | 5322519 | 60 | |
| BC057011.1 | - | 5 - 6 | 562 | 60 | ||
| BC071228.1 | - | 6 - 7 | 912 | 4915902 | 60 | |
| X76858.1 | - | 5 - 6 | 695 | 60 | ||
More Information
Set Membership: |
|

E4f1
Genomic Map