Assay Details
| Gene Symbol: Msrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013759.2 | NP_038787.1 | 2 - 3 | 274 | 72 | |
| GenBank mRNA | AF195142.1 | - | 2 - 3 | 249 | 72 | |
| AK002339.1 | - | 2 - 3 | 234 | 72 | ||
| AK002652.1 | - | 2 - 3 | 233 | 72 | ||
| AK003112.1 | - | 2 - 3 | 234 | 72 | ||
| AK003624.1 | - | 2 - 3 | 234 | 72 | ||
| BC090646.1 | - | 2 - 3 | 274 | 30946285 | 72 | |
| BC141146.1 | - | 2 - 3 | 238 | 9055758 | 72 | |
| CX566836.1 | - | 2 - 3 | 274 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msrb1
Genomic Map