Assay Details
| Gene Symbol: Msrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013759.2 | NP_038787.1 | 4 - 5 | 417 | 130 | |
| GenBank mRNA | AF195142.1 | - | 4 - 5 | 392 | 130 | |
| AK002339.1 | - | 4 - 5 | 377 | 130 | ||
| AK002652.1 | - | 4 - 5 | 376 | 130 | ||
| AK003624.1 | - | 4 - 5 | 377 | 130 | ||
| BC090646.1 | - | 4 - 5 | 417 | 30946285 | 130 | |
| BC141146.1 | - | 4 - 5 | 381 | 9055758 | 130 | |
| CX566836.1 | - | 4 - 5 | 417 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
response to oxidative stress
actin filament polymerization
protein repair
innate immune response
oxidation-reduction process
actin binding
peptide-methionine (S)-S-oxide reductase activity
zinc ion binding
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor
peptide-methionine (R)-S-oxide reductase activity
metal ion binding
methionine-R-sulfoxide reductase activity
immune system process
response to oxidative stress
actin filament polymerization
protein repair
innate immune response
oxidation-reduction process
actin binding
peptide-methionine (S)-S-oxide reductase activity
zinc ion binding
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor
peptide-methionine (R)-S-oxide reductase activity
metal ion binding
methionine-R-sulfoxide reductase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064299 | AK003112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990919 | NM_013759.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990919 | AF195142.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 965585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190018 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61173 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179825 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990919 | NM_013759.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990919 | AF195142.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064299 | AK003112.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393634 | NM_013759.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393634 | AF195142.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31099296 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03294000 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179825 | - | - |
| Whole Mouse Genome G4122A | A_51_P393634 | NM_013759.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393634 | AF195142.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138702 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179825 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13415578 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13415576 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13415565 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13415567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31099343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13415533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13415558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13510609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13066905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13456597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13205667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13205669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13415552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13205668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13066903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13415554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31099291 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442599 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442597 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418888_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442596 | - | - |
| Mouse Genome 430 2.0 Array | 1418888_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1418888_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27361_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27361_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700002748.mm.1 | NM_013759.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002748.mm.1 | TC1700000351.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700002749.mm.1 | NM_013759.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700002749.mm.1 | TC1700000351.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418888_PM_a_at | - | - |
| Mouse Expression Set 430 | 1418888_a_at | - | - |
| Murine Genome U74v2 Set | 160275_at | - | - |
| Murine Genome U74v2 Set | 160275_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418888_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418888_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418888_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418888_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3710041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3710041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5270403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003710041 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1238853 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257142 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238853 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257142 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001732.1_22-S | - | - |
| mouse-6 v1.1 (Illumina) | scl51004.4_55-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3710041 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51004.4_55-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001732.1_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710041 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257142 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2755534 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2755534 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257142 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl51004.4_55-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001732.1_22-S | - | - |

Msrb1
Genomic Map