Assay Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080773.2 | NP_001074242.1 | 8 - 9 | 995 | 142 | |
| NM_001286662.1 | NP_001273591.1 | 8 - 9 | 913 | 142 | ||
| NM_011062.4 | NP_035192.2 | 8 - 9 | 995 | 142 | ||
| GenBank mRNA | AF086625.1 | - | 8 - 9 | 961 | 142 | |
| AF126294.1 | - | 8 - 9 | 936 | 142 | ||
| AK147149.1 | - | 8 - 9 | 987 | 142 | ||
| AK147835.1 | - | 8 - 9 | 970 | 142 | ||
| AK149162.1 | - | 8 - 9 | 929 | 142 | ||
| AK149289.1 | - | 8 - 9 | 953 | 142 | ||
| AK162677.1 | - | 8 - 9 | 955 | 142 | ||
| AK165564.1 | - | 8 - 9 | 932 | 142 | ||
| AY062008.1 | - | 8 - 9 | 891 | 142 | ||
| BC146001.1 | - | 8 - 9 | 932 | 40131304 | 142 | |
| BC146003.1 | - | 8 - 9 | 932 | 40131306 | 142 | |
More Information
Set Membership: |
|

Pdpk1
Genomic Map