Assay Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080773.2 | NP_001074242.1 | 9 - 10 | 1090 | 158 | |
| NM_001286662.1 | NP_001273591.1 | 9 - 10 | 1008 | 158 | ||
| NM_011062.4 | NP_035192.2 | 9 - 10 | 1090 | 158 | ||
| GenBank mRNA | AF086625.1 | - | 9 - 10 | 1056 | 158 | |
| AF126294.1 | - | 9 - 10 | 1031 | 158 | ||
| AK147149.1 | - | 9 - 10 | 1082 | 158 | ||
| AK147835.1 | - | 9 - 10 | 1065 | 158 | ||
| AK149162.1 | - | 9 - 10 | 1024 | 158 | ||
| AK149289.1 | - | 9 - 10 | 1048 | 158 | ||
| AK162677.1 | - | 9 - 10 | 1050 | 158 | ||
| AK165564.1 | - | 9 - 10 | 1027 | 158 | ||
| AY062008.1 | - | 9 - 10 | 986 | 158 | ||
| BC146001.1 | - | 9 - 10 | 1027 | 40131304 | 158 | |
| BC146003.1 | - | 9 - 10 | 1027 | 40131306 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdpk1
Genomic Map