Assay Details
| Gene Symbol: Eci1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010023.4 | NP_034153.2 | 2 - 3 | 147 | 84 | |
| GenBank mRNA | AK004838.1 | - | 2 - 3 | 148 | 84 | |
| AK029481.1 | - | 2 - 3 | 148 | 84 | ||
| BC022712.1 | - | 2 - 3 | 131 | 4487371 | 84 | |
| BC054444.1 | - | 2 - 3 | 134 | 6476161 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eci1
Genomic Map