Assay Details
| Gene Symbol: Nthl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008743.2 | NP_032769.2 | 1 - 2 | 168 | 75 | |
| GenBank mRNA | AB006812.1 | - | 1 - 2 | 114 | 75 | |
| BC138852.1 | - | 1 - 2 | 131 | 8861874 | 75 | |
| BC138855.1 | - | 1 - 2 | 131 | 8861877 | 75 | |
| CX565655.1 | - | 1 - 2 | 147 | 75 | ||
| Y09688.1 | - | 1 - 2 | 140 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nthl1
Genomic Map