Assay Details
| Gene Symbol: Caskin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027937.2 | NP_082213.2 | 11 - 12 | 1341 | 61 | |
| GenBank mRNA | AK129327.1 | - | 11 - 12 | 1117 | 61 | |
| BC059916.1 | - | 2 - 3 | 173 | 61 | ||
| BC060720.1 | - | 11 - 12 | 1105 | 61 | ||
| BC138443.1 | - | 11 - 12 | 1249 | 8861463 | 61 | |
| BC145281.1 | - | 11 - 12 | 1249 | 9053835 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Caskin1
Genomic Map