Assay Details
| Gene Symbol: Caskin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027937.2 | NP_082213.2 | 15 - 16 | 1725 | 68 | |
| GenBank mRNA | BC059916.1 | - | 6 - 7 | 557 | 68 | |
| BC060720.1 | - | 15 - 16 | 1489 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Caskin1
Genomic Map