Assay Details
| Gene Symbol: Rab26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177375.1 | NP_796349.1 | 5 - 6 | 646 | 65 | |
| GenBank mRNA | AB232621.2 | - | 5 - 6 | 482 | 65 | |
| BC094931.1 | - | 5 - 6 | 554 | 6436242 | 65 | |
| BC118018.1 | - | 5 - 6 | 787 | 40103722 | 65 | |
More Information
Set Membership: |
|

Rab26
Genomic Map