Assay Details
| Gene Symbol: Rab26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177375.1 | NP_796349.1 | 6 - 7 | 709 | 95 | |
| GenBank mRNA | AB232621.2 | - | 6 - 7 | 545 | 95 | |
| AK043912.1 | - | 5 - 6 | 944 | 95 | ||
| BC094931.1 | - | 6 - 7 | 617 | 6436242 | 95 | |
| BC118018.1 | - | 6 - 7 | 850 | 40103722 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab26
Genomic Map