Assay Details
| Gene Symbol: Rab26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177375.1 | NP_796349.1 | 8 - 9 | 851 | 66 | |
| GenBank mRNA | AB232621.2 | - | 8 - 9 | 687 | 66 | |
| AK043912.1 | - | 7 - 8 | 1086 | 66 | ||
| BC094931.1 | - | 8 - 9 | 759 | 6436242 | 66 | |
| BC118018.1 | - | 8 - 9 | 992 | 40103722 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transport
intracellular protein transport
vesicle docking involved in exocytosis
small GTPase mediated signal transduction
protein secretion
protein transport
regulation of exocytosis
Rab protein signal transduction
exocrine system development
Golgi to plasma membrane protein transport
regulated exocytosis
protein localization to plasma membrane
nucleotide binding
GTPase activity
protein binding
GTP binding
GMP binding
transport
intracellular protein transport
vesicle docking involved in exocytosis
small GTPase mediated signal transduction
protein secretion
protein transport
regulation of exocytosis
Rab protein signal transduction
exocrine system development
Golgi to plasma membrane protein transport
regulated exocytosis
protein localization to plasma membrane
nucleotide binding
GTPase activity
protein binding
GTP binding
GMP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085370 | AK080607.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085370 | NM_001172114.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085370 | NM_153792.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085370 | NM_001172113.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P448870 | AK043912.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P448870 | BC094931.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P448870 | NM_177375.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P448870 | AK043912.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P448870 | BC094931.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P448870 | NM_177375.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655753 | D85818.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655753 | NM_011293.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195387 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136966 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159958 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P448870 | AK043912.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P448870 | BC094931.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P448870 | NM_177375.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P448870 | AK043912.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P448870 | BC094931.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P448870 | NM_177375.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P179342 | AK080607.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P448870 | AK043912.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P448870 | BC094931.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P448870 | NM_177375.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P448870 | AK043912.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P448870 | BC094931.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P448870 | NM_177375.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13324380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31097979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13540383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13324374 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10448623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448621 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448616 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448620 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448619 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448629 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10448630 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435500_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10448615 | - | - |
| Mouse Genome 430 2.0 Array | 1435500_at | - | - |
| Mouse Genome 430 2.0 Array | 1435500_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 328778_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 328778_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700014551.mm.1 | NM_177375.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014551.mm.1 | NM_177375.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014551.mm.1 | TC1700002798.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700014552.mm.1 | NM_177375.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014552.mm.1 | NM_177375.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014552.mm.1 | TC1700002798.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700014553.mm.1 | NM_177375.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014553.mm.1 | NM_177375.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014553.mm.1 | TC1700002798.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700014554.mm.1 | NM_177375.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014554.mm.1 | NM_177375.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700014554.mm.1 | TC1700002798.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435500_at | - | - |
| Mouse Expression Set 430 | 1435500_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435500_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435500_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 619090 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2722058 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2722058 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2718406 | - | - |

Rab26
Genomic Map