Assay Details
| Gene Symbol: Triobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024716.1 | NP_001019887.1 | 3 - 4 | 274 | 109 | |
| NM_001039156.1 | NP_001034245.1 | 14 - 15 | 5041 | 109 | ||
| NM_138579.4 | NP_613045.3 | 13 - 14 | 4903 | 109 | ||
| GenBank mRNA | AK053194.1 | - | 3 - 4 | 234 | 109 | |
| AK085512.1 | - | 3 - 4 | 244 | 109 | ||
| AK129417.1 | - | 3 - 4 | 224 | 109 | ||
| AK170390.1 | - | 3 - 4 | 220 | 109 | ||
| BC003984.1 | - | 1 - 2 | 70 | 109 | ||
| BC042760.1 | - | 3 - 4 | 213 | 109 | ||
| BC053329.1 | - | 3 - 4 | 258 | 109 | ||
| CF533172.1 | - | 3 - 4 | 346 | 109 | ||
| DQ228000.1 | - | 14 - 15 | 5041 | 109 | ||
| DQ228001.2 | - | 13 - 14 | 4903 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Triobp
Genomic Map