Assay Details
| Gene Symbol: Ddx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152806.3 | NP_690019.2 | 1 - 2 | 1249 | 108 | |
| NM_199080.2 | NP_951062.1 | 12 - 13 | 1953 | 108 | ||
| GenBank mRNA | AK027954.1 | - | 1 - 2 | 99 | 108 | |
| AK151061.1 | - | 12 - 13 | 1728 | 108 | ||
| AK152749.1 | - | 12 - 13 | 1729 | 108 | ||
| AK152831.1 | - | 12 - 13 | 1728 | 108 | ||
| AK152910.1 | - | 1 - 2 | 188 | 108 | ||
| BC027758.1 | - | 1 - 2 | 486 | 108 | ||
| BC038378.1 | - | 1 - 2 | 835 | 108 | ||
| BI655302.1 | - | 5 - 6 | 625 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK027954.1 transcripts.
Set Membership: |
|

Ddx17
Genomic Map