Assay Details
| Gene Symbol: Ddx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040187.1 | NP_001035277.1 | 8 - 9 | 1480 | 93 | |
| NM_199079.1 | NP_951061.1 | 8 - 9 | 1299 | 93 | ||
| NM_199080.2 | NP_951062.1 | 8 - 9 | 1480 | 93 | ||
| GenBank mRNA | AK039888.1 | - | 8 - 9 | 1299 | 93 | |
| AK151061.1 | - | 8 - 9 | 1255 | 93 | ||
| AK152749.1 | - | 8 - 9 | 1256 | 93 | ||
| AK152831.1 | - | 8 - 9 | 1255 | 93 | ||
| AK161001.1 | - | 8 - 9 | 1297 | 93 | ||
| AK161012.1 | - | 8 - 9 | 1295 | 93 | ||
| BC062910.1 | - | 8 - 9 | 1285 | 6816663 | 93 | |
| BC096036.1 | - | 8 - 9 | 1480 | 30549388 | 93 | |
| BI655302.1 | - | 1 - 2 | 152 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx17
Genomic Map