Assay Details
| Gene Symbol: Ddx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040187.1 | NP_001035277.1 | 9 - 10 | 1587 | 69 | |
| NM_199079.1 | NP_951061.1 | 9 - 10 | 1406 | 69 | ||
| NM_199080.2 | NP_951062.1 | 9 - 10 | 1587 | 69 | ||
| GenBank mRNA | AK039888.1 | - | 9 - 10 | 1406 | 69 | |
| AK151061.1 | - | 9 - 10 | 1362 | 69 | ||
| AK152749.1 | - | 9 - 10 | 1363 | 69 | ||
| AK152831.1 | - | 9 - 10 | 1362 | 69 | ||
| AK161001.1 | - | 9 - 10 | 1404 | 69 | ||
| AK161012.1 | - | 9 - 10 | 1402 | 69 | ||
| BC096036.1 | - | 9 - 10 | 1587 | 30549388 | 69 | |
| BI655302.1 | - | 2 - 3 | 259 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx17
Genomic Map