Assay Details
| Gene Symbol: Pick1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045558.1 | NP_001039023.1 | 10 - 11 | 1028 | 76 | |
| NM_008837.3 | NP_032863.2 | 10 - 11 | 1087 | 76 | ||
| GenBank mRNA | AK050224.1 | - | 7 - 8 | 1853 | 76 | |
| AK090155.1 | - | 1020 | 76 | |||
| BC023088.1 | - | 9 - 10 | 1623 | 76 | ||
| BC030363.1 | - | 2 - 3 | 1332 | 76 | ||
| BC030880.1 | - | 1 - 2 | 1163 | 76 | ||
| BC030912.1 | - | 1 - 2 | 239 | 76 | ||
| BC040824.1 | - | 1 - 2 | 1163 | 76 | ||
| BC048788.1 | - | 10 - 11 | 973 | 6488048 | 76 | |
| BE652516.1 | - | 2 - 3 | 223 | 76 | ||
| CD363874.1 | - | 7 - 8 | 517 | 76 | ||
| Z46720.1 | - | 9 - 10 | 776 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pick1
Genomic Map